Orders LinkContactUsLinkComponent
Dissection boards, DissecTable™
Dissection boards, DissecTable™
Board base, Blue, 481×656×91 mm
Catalog # 720-1686
Supplier:   Simport Scientific
Dissection boards, DissecTable™
Catalog # 720-1686
Supplier:  Simport Scientific
Dissection boards, DissecTable™
Dissection boards, DissecTable™
Board base, Blue, 481×656×91 mm
Price
311,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
HDPE / PS. These dissection boards feature a large cutting area on one side. They include dimensional scales in inches and centimeters, a grid and dimensional circles of different sizes. The other side offers two cutting boards half the size with the same dimensional features printed on each one of them.
  • Drain groove is carved all around the edge of the boards in order to contain fluids
  • All corners have rubber feet giving more stability to the working surface
  • Bases are stackable and will not move sideways during dissecting work
Specifications
  • Colour:
    Blue
  • Description:
    Board base
  • W×D×H:
    481×656×91 mm