Order Entry
Belgium
Orders LinkContactUsLinkComponent
Eyewash solution, pH neutral

Eyewash solution, pH neutral

Eyewash solution, pH neutral, 200 ml bottle
Catalog # 118-1707
Supplier:   Plum Safety
Eyewash solution, pH neutral
Catalog # 118-1707
Supplier:  Plum Safety
Eyewash solution, pH neutral
Eyewash solution, pH neutral

Eyewash solution, pH neutral, 200 ml bottle

Price
17,10 €
Log in for availability
1 * 1 ST
Product Details & Documents

Sterile phosphate buffer solution (4,9%) for fast and effective neutralisation of acids and alkalis.

  • Handy sized bottle, well suited for use in first aid kits and emergency bags
  • Bottle with ergonomic eye cup, dust cap and clear instructions
  • Can be used as refill for Plum wall-mounted eyewash station
  • No maintenance and cleaning of the bottles required
  • Bottles in single carton: Carton protects the bottles from external contaminations and can be opened quickly and easily at the perforation

Rinsing time: ~2 minutes per 200 ml
Shelf life: 3 years

According to DIN EN 15154-4

Certifications: According to DIN EN 15154-4.

Specifications
  • Description:
    Eyewash solution, pH neutral, 200 ml bottle
  • Seq:
    01
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Belt bag for 200 ml eyewash bottles
    Belt bag for 200 ml eyewash bottles
    118-1704
    Log in for availability
    1 * 1 ST - 9,90 €

Product Information

  • Catalog #
    Availability
    Price
  • Blue wall-mountable holder for 200 ml pH neutral eyewash bottles
    Blue wall-mountable holder for 200 ml pH neutral eyewash bottles
    118-1712
    Log in for availability
    1 * 1 ST - 5,10 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.