Order Entry
Belgium
Orders LinkContactUsLinkComponent
Phosphate Buffer, pH 6.6 at 25 °C

Phosphate Buffer, pH 6.6 at 25 °C

Phosphate buffer, 6,6, 1 vial in poly bottle, 1
Catalog # P8165-1VL
Phosphate Buffer, pH 6.6 at 25 °C
Catalog # P8165-1VL
CAS Number:  
Phosphate Buffer, pH 6.6 at 25 °C

Phosphate buffer, 6,6, 1 vial in poly bottle, 1

CAS Number:  
Price
115,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Phosphate buffer's buffering capacity is pH sensitive and is preferred to be used at physiological pH (7,2 to 7,4).

  • Each vial will prepare 3,8 L of buffer, 34 mM
  • Used in hematology and histology

For use with Wright Stain for Geometric Data HEMASTAIN®. Phosphate buffer serves as an important fixative for light and electron microscopy. It is the most commonly used component of irrigation solution during surgeries, to preserve living tissues. It is also used in immunochemical staining procedures.

Documents

Specifications

Specifications

  • Pack type:
    1 vial in poly bottle
  • Pk:
    1
  • Description:
    Phosphate buffer
  • pH value:
    6,6
  • Cat. No.:
    P8165-1VL
  • M-Clarity quality level:
    MQ200
  • Storage temperature:
    RT
  • Appearance:
    White powder

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.