Percoll®, Siam
Percoll®
Catalog # P1644-25ML
Supplier: SIGMA ALDRICH MICROSCOPY
Jump to:
Product Details & Documents
Percoll® is a classic medium for density gradient centrifugation of cells, viruses, and subcellular particles. It consists of colloidal silica particles of 15 – 30 nm diameter (23% w/w in water), coated with polyvinylpyrrolidone (PVP).
- Non toxic
- Ideal for use with biological materials
Documents
Certificates
Search by Lot NumberSpecifications
- Désignation:Percoll®
- Seq:02