MALACHITE GREEN SOLUTION 1 * 500 mL
Catalog # HT80216-500ML
Supplier: SIGMA ALDRICH MICROSCOPY
Frequently Bought Together
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Potassium hydroxide, pellets, pure, Supelco®
Supplier: Merck
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
Reference substances for GC analysis, Supelco®
Supplier: Merck
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
NANOCOLOR® Photometric Tube Tests, Total Nitrogen
Supplier: MACHEREY-NAGEL
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Colour comparison disk Ammonium suitable for Cat. No. 920006 VISOCOLOR HE. 1 * 1 items
Supplier: MACHEREY-NAGEL
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
GLYCOL REAGENT 3 1 * 25 items
Supplier: Hach
Methyl-tert.butylether CAS:1634-04-4 EC:216-653-1 1 * 5 mL
Supplier: CUSTOM MADE CHEMICALS LAB