Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffers, colour coded, reference standards, Supelco®

pH buffers, colour coded, reference standards, Supelco®

4,00, ±0,01, Red, 25 °C, Plastic container with spigot, 1×gallon, 1 gal US
Catalog # B5020-1GA
Supplier:   Merck
pH buffers, colour coded, reference standards, Supelco®
Catalog # B5020-1GA
Supplier:  Merck
CAS Number:  
pH buffers, colour coded, reference standards, Supelco®
pH buffers, colour coded, reference standards, Supelco®

4,00, ±0,01, Red, 25 °C, Plastic container with spigot, 1×gallon, 1 gal US

CAS Number:  
Price
233,00 €
Log in for availability
1 * 1 gal us
Product Details & Documents

Certifications: Standardised against N.I.S.T. standard reference materials

Documents

Specifications

Specifications

  • Pack type:
    Plastic container with spigot
  • Pk:
    1 gal US
  • Reference temperature:
    25 °C
  • Colour:
    Red
  • pH accuracy:
    ±0,01
  • Pk Info:
    1×gallon
  • pH value:
    4,00
  • Cat. No.:
    B5020-1GA
  • M-Clarity quality level:
    MQ100

Specifications

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.