Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffers, colour coded, reference standards, Supelco®

pH buffers, colour coded, reference standards, Supelco®

7,00, ±0,01, Yellow, 25 °C, Plastic bottle, 1×500 ml, 500 mL
N° de catalogue B4770-500ML
Fournisseur:   Merck
pH buffers, colour coded, reference standards, Supelco®
N° de catalogue B4770-500ML
Fournisseur:  Merck
Numéro CAS:  
pH buffers, colour coded, reference standards, Supelco®
pH buffers, colour coded, reference standards, Supelco®

7,00, ±0,01, Yellow, 25 °C, Plastic bottle, 1×500 ml, 500 mL

Numéro CAS:  
Prix
49,60 €
Log in for availability
1 * 500 mL
Product Details & Documents

Certifications: Standardised against N.I.S.T. standard reference materials

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Colour:
    Yellow
  • pH accuracy:
    ±0,01
  • Pk Info:
    1×500 ml
  • pH value:
    7,00
  • Cat. No.:
    B4770-500ML
  • M-Clarity quality level:
    MQ100

Specifications