Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffers, colour coded, reference standards, Supelco®

pH buffers, colour coded, reference standards, Supelco®

7,00, ±0,01, Yellow, 25 °C, Plastic container with spigot, 1×gallon, 1 gal US
Catalog # B4770-1GA
Supplier:   Merck
pH buffers, colour coded, reference standards, Supelco®
Catalog # B4770-1GA
Supplier:  Merck
CAS Number:  
pH buffers, colour coded, reference standards, Supelco®
pH buffers, colour coded, reference standards, Supelco®

7,00, ±0,01, Yellow, 25 °C, Plastic container with spigot, 1×gallon, 1 gal US

CAS Number:  
Price
259,00 €
Log in for availability
1 * 1 gal us
Product Details & Documents

Certifications: Standardised against N.I.S.T. standard reference materials

Documents

Specifications

Specifications

  • Pack type:
    Plastic container with spigot
  • Pk:
    1 gal US
  • Reference temperature:
    25 °C
  • Colour:
    Yellow
  • pH accuracy:
    ±0,01
  • Pk Info:
    1×gallon
  • pH value:
    7,00
  • Cat. No.:
    B4770-1GA
  • M-Clarity quality level:
    MQ100

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.