Orders LinkContactUsLinkComponent
Dissection boards, disposable
Dissection boards, disposable
Disposable dissection boards, White, 350×333 mm
Catalog # LPIT197032
Supplier:   LP ITALIANA
Dissection boards, disposable
Catalog # LPIT197032
Supplier:  LP ITALIANA
Dissection boards, disposable
Dissection boards, disposable
Disposable dissection boards, White, 350×333 mm
Price
322,00 €
Log in for availability
1 * 24 ST
Product Details & Documents
Disposable cutting boards made of cardboard and PS foam for the dissection of surgical specimens.
  • With data registration area
  • Pre-printed scale (in mm)
  • Three different sizes available
Specifications
  • Colour:
    White
  • Description:
    Disposable dissection boards
  • L×W:
    350×333 mm
  • Seq:
    03