Pipette tips
Bevelled, non-graduated, 1 - 200 µl, +, 50 mm, Yellow, 96 tips/rack; 10 racks/case
Catalog # ALPCLW6365S
Supplier: Alpha Laboratories
Jump to:
Product Details & Documents
These tips are manufactured from medical grade PP in modern production facilities with stringent quality controls at every stage.
- Tips are lot tested and certified for absence of detectable human DNA and detectable endotoxins (pyrogens)
- Tips are tested and certified for absence of heavy metals, detectable DNase and RNase
- With graduations to visually check volume
- Suitable for all major pipettor brands
Specifications
- Colour code:Yellow
- Packed:96 tips/rack; 10 racks/case
- Sterile:+
- Volume:1 - 200 µl
- Length:50 mm
- Seq:05
- Tip style:Bevelled, non-graduated