Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Tissue processing cassette, System II Hex

Blue
  720-1893
 :   CELLPATH
Tissue processing cassette, System II Hex
  720-1893
 :  CELLPATH
Tissue processing cassette, System II Hex
Tissue processing cassette, System II Hex

Blue

 
68,50 €
   
1 * 1.000 ST
 

The System II Hex Cassette differs from conventional round hole processing cassettes in that it has hexagonal instead of round perforations arranged in a honeycombed structure to better utilise the available surface area.

  • Available in different stock colours, which allow laboratories to colour code different types of workload or different applications
  • Manufactured from high–grade virgin acetyl resin

This unique arrangement increases the open area by approximately 13 percent thereby increasing reagent flow through and improving specimen staining and analysis time.

A potential cost saving benefit is that there is less carry over of chemicals so reducing the frequency of chemical change.

Although made to an international industry standard size, the Hex Cassette is designed to suit all established cassette–writing machines and benefits from extra thick side walls, so increasing stability and rigidity, which is crucial for high quality sectioning.

 
  • Colour:
    Blue