Labels, hazard warning
Hazard labels: gas cylinder, PIC 335-TRI 100-B7541, Polyester, Black on yellow, 100×87 mm, 3 labels/card
Catalog # BRDY250302
Supplier: Brady
Jump to:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specifications
- Colour:Black on yellow
- Description:Hazard labels: gas cylinder
- Dimensions:100×87 mm
- Material:Polyester
- Seq:31
- Model:PIC 335-TRI 100-B7541
- Packed:3 labels/card
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



