Orders LinkContactUsLinkComponent
Pycnometer
5 ml
Catalog # HECH42571008
Pycnometer
Catalog # HECH42571008
Pycnometer
Pycnometer
5 ml
Price
69,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
  • With ground-in stopper
  • Good chemical resistance
  • Good temperature resistance
 : DIN ISO 3507
Specifications
  • Volume (nom.):
    5 ml