Orders LinkContactUsLinkComponent
Cylinders, without lid
Cylinders, without lid
80 mm
Catalog # HECH44409018
Cylinders, without lid
Catalog # HECH44409018
Cylinders, without lid
Cylinders, without lid
80 mm
Price
11,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
Jars, made of glass or stainless steel, with base, without lids.
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Ordering information: Lids with knob available must be ordered separately.
Specifications
  • Ext.Ø:
    80 mm
  • Seq:
    17
  • Cat. No.:
    HECH44409018