Order Entry
Belgium
Orders LinkContactUsLinkComponent
Applicators

Applicators

Sterile swabs, Aluminium/cotton, 150 mm, 0,9 – 1,0 mm
Catalog # HERE1032749
Supplier:   HERENZ HEINZ
Applicators
Catalog # HERE1032749
Supplier:  HERENZ HEINZ
Applicators
Applicators

Sterile swabs, Aluminium/cotton, 150 mm, 0,9 – 1,0 mm

Price
85,60 €
Log in for availability
1 * 100 ST
Product Details & Documents

Aluminium applicators with a single cotton bud.

Packaging: In a single peel-pack.

Specifications
  • Ø:
    0,9 – 1,0 mm
  • Description:
    Sterile swabs
  • Length:
    150 mm
  • Shaft/head:
    Aluminium/cotton