Orders LinkContactUsLinkComponent
Micro pipettes, disposable, with ring mark
Micro pipettes, disposable, with ring mark
50 µl, Green, One mark
Catalog # HECH40555021
Micro pipettes, disposable, with ring mark
Catalog # HECH40555021
Micro pipettes, disposable, with ring mark
Micro pipettes, disposable, with ring mark
50 µl, Green, One mark
Price
31,80 €
Log in for availability
1 * 250 ST
Product Details & Documents
Disposable micro pipettes with ring mark.
  • Calibrated to contain 'In'
  • Colour-coded
 : DIN ISO 7550
Specifications
  • Colour code:
    Green
  • Type:
    One mark
  • Capacity:
    50 µl
  • Seq:
    04