Orders LinkContactUsLinkComponent
Wire baskets, square
Wire baskets, square
160×160×160 mm
Catalog # HECH41075016
Wire baskets, square
Catalog # HECH41075016
Wire baskets, square
Wire baskets, square
160×160×160 mm
Price
143,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These square baskets are made of stainless steel. Designed for transporting, storing, cleaning and sterilising laboratory utensils.
  • Excellent temperature resistance
  • Good chemical resistance
  • Reinforced edges
Specifications
  • W×D×H:
    160×160×160 mm