Orders LinkContactUsLinkComponent
Watchmaker forceps
Watchmaker forceps
Watchmaker No. 3, straight, 115 mm
Catalog # 232-0201
Supplier:   MURRAY S
Watchmaker forceps
Catalog # 232-0201
Supplier:  MURRAY S
Watchmaker forceps
Watchmaker forceps
Watchmaker No. 3, straight, 115 mm
Price
12,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel. Ideal for small-scale dissection applications or precise part placement.
  • Very fine points
  • Smooth flat handle
  • Available straight or curved
Specifications
  • Type:
    Watchmaker No. 3, straight
  • Length:
    115 mm