Orders LinkContactUsLinkComponent
Inserts for screw neck vials
Inserts for screw neck vials
Low volume, with poly spring, 0,15 ml, Glass, No
Catalog # WATEWAT094171
Marketsource
Supplier:   WATERS
Inserts for screw neck vials
Catalog # WATEWAT094171
Supplier:  WATERS
Marketsource
Inserts for screw neck vials
Low volume, with poly spring, 0,15 ml, Glass, No
Price
97,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Specifications
  • Cap included:
    No
  • Capacity:
    0,15 ml
  • Description:
    Low volume, with poly spring
  • Material:
    Glass
  • Seq:
    02