ContactUsLinkComponent
Vial, capacity 2ml, target lovial, 11mm crimp top, amber, silanized glass I-D™, national scientific silanized, diameter 12mm, height 32mm, flat bottom, patched, vial surface is deactivated, residual volume <170µl
Catalog # CRMAC4011-S2W
Supplier:  CHROMACOL
Vial, capacity 2ml, target lovial, 11mm crimp top, amber, silanized glass I-D™, national scientific silanized, diameter 12mm, height 32mm, flat bottom, patched, vial surface is deactivated, residual volume <170µl
Catalog # CRMAC4011-S2W
Supplier:  CHROMACOL