Vial, capacity 2ml, target 10-425 wide opening screw thread, diameter 12mm, height 32mm, flat bottom, material glass, colour amber, usable volume 1.5ml, compatible with many non-robotic autosamplers
Catalog # CRMAC4010-2
Supplier: CHROMACOL