Vial, capacity 4ml, colour amber, material glass, 13-425 screw thread, flat bottom, patched, usable volume 4ml, diameter 15mm, heigh t 45mm, residual volume <800µl, frequently used with waters WISP autosamplers
Catalog # CRMAC4015-2W
Supplier: CHROMACOL