ContactUsLinkComponent
Vial, capacity 2ml, target DP screw thread, colour clear, silanized glass I-D, diameter 12mm, height 32mm, vial finish 9mm screw thr ead, flat bottom, usable volume 1.5ml, residual volume <170µl
Catalog # CRMAC4000-S1W
Supplier:  CHROMACOL
Vial, capacity 2ml, target DP screw thread, colour clear, silanized glass I-D, diameter 12mm, height 32mm, vial finish 9mm screw thr ead, flat bottom, usable volume 1.5ml, residual volume <170µl
Catalog # CRMAC4000-S1W
Supplier:  CHROMACOL