Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Cork stoppers

26 mm, 19 mm, 23 mm
Catalog # HECH41594023
Cork stoppers
Catalog # HECH41594023
Cork stoppers
Cork stoppers

26 mm, 19 mm, 23 mm

Price
407,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Cork stoppers designed for medical applications.

Specifications
  • Ø bottom:
    19 mm
  • Ø top:
    23 mm
  • Height:
    26 mm