Order Entry
Belgium
Orders LinkContactUsLinkComponent
Automatic tilt measures

Automatic tilt measures

Amyl alcohol, 1 ml, Clear
Catalog # 614-0148
Supplier:   GERBER FUNKE
Automatic tilt measures
Catalog # 614-0148
Supplier:  GERBER FUNKE
Automatic tilt measures
Automatic tilt measures

Amyl alcohol, 1 ml, Clear

Price
60,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

Glass, with rubber stopper.

  • Superior quality models
  • Storage bottle of 500/250 ml
  • For milk analysis applications

Colour: Clear

Specifications
  • Capacity:
    1 ml
  • Colour:
    Clear
  • Description:
    Amyl alcohol
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.