MULTIPLE STANDARD SOL. LOW VARION ES/1 1 * 1 L
WTWO107050
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
pH buffers, ready to use
: WTW
VWR®, Redox (ORP) Standards
: VWR Chemicals
Buffer solutions
: REAGECON
Hydrochloric acid 0.01 mol/l (0.01 N) Reag. Ph. Eur. 1043504
: VWR Chemicals
pH buffer solutions
: Mettler - Toledo
REDOX BUFFER 475 MV, 1 * 500 mL
: HAMILTON BONADUZ
pH buffers, DuraCal
: HAMILTON BONADUZ
VWR®, COD calibration standards
: VWR Chemicals
Dicamba certified 1 * 0,25 g
: EHRENSTORFER
2,2,2-Trichloroethanol
: EHRENSTORFER
Etridiazole certified 1 * 0,1 g
: EHRENSTORFER