Order Entry
Belgium
Orders LinkContactUsLinkComponent
Proteinase K solution

Proteinase K solution

10 mL
Catalog # A4392.0010
Supplier:   PanReac AppliChem
Proteinase K solution
Catalog # A4392.0010
Supplier:  PanReac AppliChem
CAS Number:  
Proteinase K solution
Proteinase K solution

10 mL

CAS Number:  
Price
226,00 €
Log in for availability
1 * 10 mL
Product Details & Documents

Proteinase K is used to destruct proteins in cell lysates (tissue, cultured cells) and to liberate nucleic acids, since it very effectively digests DNases and RNases.

  • DNases/RNases not detectable
  • Proteinase K min. 600 mAnsonU/ml
  • GHS symbols: GHS08
  • GHS signal word: Danger

Composition:
CaCl₂ · 2H₂O 1 mM
Glycerol (v/v) 40%
Proteinase K 20 mg/ml
Tris · HCl (pH 7,5) 10 mM

Specifications

Specifications

  • Pk:
    10 mL
  • Cat. No.:
    A4392.0010
  • Storage conditions:
    2...8 °C
  • Enzyme synonyms:
    Protease K solution 20 mg/ml, Endopeptidase K solution 20 mg/ml, Tritirachium alkaline proteinase solution 20 mg/ml, Tritirachium album serine proteinase solution 20 mg/ml, Peptidase K solution 20 mg/ml
  • Concentration:
    20 mg/ml
  • Enzyme name:
    Proteinase K solution

Specifications