ContactUsLinkComponent
Refrigerator ct2041 total capacity-195l with freezer volume-44l dimensions-height-121.5 cm width-55.2 cm depth-61.3 cm energy-a stai nless s teel freezer on top freestanding 1 * 1 items
Catalog # LIEBCT2041
Supplier:  Liebherr
Refrigerator ct2041 total capacity-195l with freezer volume-44l dimensions-height-121.5 cm width-55.2 cm depth-61.3 cm energy-a stai nless s teel freezer on top freestanding 1 * 1 items
Catalog # LIEBCT2041
Supplier:  Liebherr
Frequently Bought Together