Orders LinkContactUsLinkComponent
Sodium deoxycholate
Glass bottle for solids, 25 g
  PIER89905
Sodium deoxycholate
  PIER89905
 :  302-95-4
Sodium deoxycholate
Sodium deoxycholate
Glass bottle for solids, 25 g
 :  302-95-4
630,00 €
1 * 25 g
 :  Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
  •  :302-95-4
  •  :C₂₄H₃₉NaO₄
  •  :MFCD00064139
  •  :414.56 g/mol
  •  :357…365 °C
  •  :Ambient
  •  :206-132-7
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g