Sodium deoxycholate
Glass bottle for solids, 5 g
N° de catalogue PIER89904
Fournisseur: Thermo Fisher Scientific
Sodium deoxycholate
N° de catalogue PIER89904
Fournisseur: Thermo Fisher Scientific
Numéro CAS: 302-95-4
Jump to:
Product Details & Documents
Synonyms: Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
- CAS Number:302-95-4
- Formula:C₂₄H₃₉NaO₄
- MDL Number:MFCD00064139
- Molecular Weight:414.56 g/mol
- Melting Pt:357…365 °C
- Storage Temperature:Ambient
- EINECS:206-132-7
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



