Forceps, precision
Straight, short fine tips, 2, 115 mm
Catalog # 232-0000
Supplier: OUTILS RUBIS
Jump to:
Product Details & Documents
Stainless steel. Forceps with very precisely machined tips. Ideal for applications that require a high degree of precision, including microscopy, anatomy, biology, microbiology, gene technology and electron microscopy.
- Acid-resistant
- Resistant against hydrogen fluoride and nitric acid
- 100% anti-magnetic
Documents
Specifications
- Type:2
- Description:Straight, short fine tips
- Length:115 mm



