Labels, hazard warning
Hazard labels: hot surface, —, Polyester, Black on yellow, 50×43 mm, 7 labels/card
Catalog # 116-0684
Supplier: Brady
Jump to:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specifications
- Colour:Black on yellow
- Description:Hazard labels: hot surface
- Dimensions:50×43 mm
- Material:Polyester
- Seq:16
- Model:—
- Packed:7 labels/card



