Orders LinkContactUsLinkComponent
Cold traps, Type KF
Cold trap, KF 29-GL-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity and 1000 ml coolent cap
  KGWA17372
 :   KGW
Cold traps, Type KF
  KGWA17372
 :  KGW
Cold traps, Type KF
Cold traps, Type KF
Cold trap, KF 29-GL-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity and 1000 ml coolent cap
643,00 €
1 * 1 ST
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents. Model KF cold traps have a GL 32 glass thread together with a screw-on cap (spout), through which the condensate can easily be poured out of the cold trap. Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
  • Easy to clean
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask made of blue coated metal or aluminium
  • Description:
    Cold trap, KF 29-GL-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity and 1000 ml coolent cap
  • Seq:
    04
Accessories
  •  
    Description
  • KGWA1733
    Plastic ring for cold trap, type KF
    1 * 1 ST - 62,50 €
  •  
    Description
  • KGWA1741
    Cooling finger S 29-GL
    1 * 1 ST - 275,00 €