Orders LinkContactUsLinkComponent
Temperature loggers, Thermo Button
Temperature loggers, Thermo Button
Key holder, yellow
Catalog # 620-3179
Supplier:   PROGES PLUS
Temperature loggers, Thermo Button
Catalog # 620-3179
Supplier:  PROGES PLUS
Temperature loggers, Thermo Button
Temperature loggers, Thermo Button
Key holder, yellow
Price
4,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Small and rugged, the Thermo Button data loggers can survive any harsh environment. They are suitable for temperature control, product tracking, transport, process analysis, HACCP.
  • Collect and record date and time
  • Programmable low and high alarm threshold with temporization parameters
  • Battery life up to ten years depending on sample rate and temperature
  • Resist to dust
Ø×H: 16×6 mm
Conform to IP 65.
Certifications: EN 12830 tested by CEMAGREF and LCIE, RTCA/DO-160 section 21, IATA (Lithium batteries), not tested for 'Food contact approval'.
Delivery information: A calibration certificate is available on request with extra charges, when ordering. The thermotrack PC software is in eight languages: French, English, German, Spanish, Portuguese, Slovenian, Japanese and Italian.
Documents
Specifications
  • Description:
    Key holder, yellow