Temperature loggers, Thermo Button
Key holder, green
620-3178
Small and rugged, the Thermo Button data loggers can survive any harsh environment. They are suitable for temperature control, product tracking, transport, process analysis, HACCP.
- Collect and record date and time
- Programmable low and high alarm threshold with temporization parameters
- Battery life up to ten years depending on sample rate and temperature
- Resist to dust
Ø×H: 16×6 mm
Conform to IP 65.
: EN 12830 tested by CEMAGREF and LCIE, RTCA/DO-160 section 21, IATA (Lithium batteries), not tested for 'Food contact approval'.
: A calibration certificate is available on request with extra charges, when ordering. The thermotrack PC software is in eight languages: French, English, German, Spanish, Portuguese, Slovenian, Japanese and Italian.
- Description:Key holder, green