Adapters, cone/screw thread, Quickfit®
29/32 NS, 18 mm, 7,0 - 8,5 mm
Catalogus # 201-0228
Leverancier: SciLabware
Ga naar:
Product Details & Documents
PYREX®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer. Ideal for securing thermometers, gas inlet tubes etc.
Specificaties
- Cone size:29/32 NS
- Hole Ø:7,0 - 8,5 mm
- Thread:18 mm
- Seq:10
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



