Orders LinkContactUsLinkComponent
UV/white light transilluminators
UV/white light transilluminators
Transilluminator, 365/302 nm UV/white light, 8 W, filter size 200×200 mm, LMW-20
Catalog # 732-4373
UV/white light transilluminators
Catalog # 732-4373
UV/white light transilluminators
UV/white light transilluminators
Transilluminator, 365/302 nm UV/white light, 8 W, filter size 200×200 mm, LMW-20
Price
2 480,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These UV/white light transilluminators house white light and UV working areas side-by-side. White light can be used for viewing Coomassie Blue stained gels, silver stained protein gels, autoradiographs and microtitre plates. The UV side can be used for viewing ethidium bromide gels.
  • Easily switch between light sources using the rocker switch on the front of the unit
  • UV side utilises UVG™ long-life filter
  • Clear UV blocking cover is hinged and can be height adjusted to provide a suitable viewing angle or to access the filter surface
W×D×H: 486×337×143 mm (height including UV blocking cover)
Weight: 9,6 kg
Specifications
  • Model:
    LMW-20
  • Description:
    Transilluminator, 365/302 nm UV/white light, 8 W, filter size 200×200 mm