UV/white light transilluminators
Transilluminator, 302 nm UV/white light, 8 W, filter size 200×200 mm, TMW-20
Catalog # 732-4371
Supplier: UVP ULTRA VIOLET PRODUCTS
Jump to:
Product Details & Documents
These UV/white light transilluminators house white light and UV working areas side-by-side. White light can be used for viewing Coomassie Blue stained gels, silver stained protein gels, autoradiographs and microtitre plates. The UV side can be used for viewing ethidium bromide gels.
- Easily switch between light sources using the rocker switch on the front of the unit
- UV side utilises UVG™ long-life filter
- Clear UV blocking cover is hinged and can be height adjusted to provide a suitable viewing angle or to access the filter surface
W×D×H: 486×337×143 mm (height including UV blocking cover)
Weight: 9,6 kg
Specifications
- Model:TMW-20
- Description:Transilluminator, 302 nm UV/white light, 8 W, filter size 200×200 mm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



