CellStor Pot™ specimen container
Catalog # 720-2321
Discontinued
Product Details & Documents
Prefilled specimen containers.
- Leak-proof lid for safe transportation and specimen storage
- With traceable badge number, safety data sheet in the packaging