Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions, NIST traceable, Orion™

pH buffer solutions, NIST traceable, Orion™

pH 4,01 buffer, colour-coded pink, 4,01, Pink, Plastic bottle, 475 mL
Catalog # 662-0770
Supplier:   Thermo Orion
pH buffer solutions, NIST traceable, Orion™
Catalog # 662-0770
Supplier:  Thermo Orion
CAS Number:  
pH buffer solutions, NIST traceable, Orion™
pH buffer solutions, NIST traceable, Orion™

pH 4,01 buffer, colour-coded pink, 4,01, Pink, Plastic bottle, 475 mL

CAS Number:  
Price
25,10 €
Log in for availability
1 * 475 mL
Product Details & Documents

Perform accurate, reproducible pH calibrations with Orion™ pH buffers.

  • NIST traceability
  • Colour-coded solutions are available for ease of use and identification

Solutions are manufactured under ISO 9000 quality standards.

The kit includes 1 pint (475 ml) bottle each of pH 4,00 buffer, pH 7,00 buffer, pH 10,01 buffer and standard pH electrode storage solution and one pH electrode storage bottle.

Delivery information: Supplied in 475 ml or 60 ml bottles, with buffer/temperature tables to ensure more precise calibration.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    475 mL
  • Colour:
    Pink
  • Description:
    pH 4,01 buffer, colour-coded pink
  • pH value:
    4,01

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.