Bomb sampler, Sub Sampler Pro
Bomb samplers, Saddle leather, 1000 ml, 430 mm, 57 mm
Catalog # SAMP5072A-1000
Supplier: Sampling Systems
Jump to:
Product Details & Documents
Essential for flammable liquids as a heavy-duty sampler.
- Metallic parts manufactured from stainless steel
- Unique double action to sample from a set distance
- Can be plunged until the assembly touches the bottom of the container
Specially designed for the environment throughout the world for both chemical and environmental sampling.
Certifications: Conforms to FDA CFR 177.2600.
Specifications
- Ø:57 mm
- Capacity:1000 ml
- Description:Bomb samplers
- Length:430 mm
- Material:Saddle leather
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



