Slide Boxes
White, PS, 100, 172×172×31 mm
Catalog # GLWAK99W
Supplier: GLW
Jump to:
Product Details & Documents
High quality slide storage boxes for microscopy slides.
Used for storing, transporting and freezing in science, industry and biotechnology.
Specifications
- No. of slides:100
- Colour:White
- Material:PS
- Seq:15
- W×D×H:172×172×31 mm
Accessories
Product Information
- Catalog #ColourMaterialAvailabilityPrice
- Divider for slides and coverslips of special sizesDivider for slides and coverslips of special sizesGLWAZK20GreyPSLog in for availability1 * 1 ST - 2,80 €
Product Information
- Catalog #ColourMaterialAvailabilityPrice
- Divider for slides and coverslips of special sizesDivider for slides and coverslips of special sizesGLWAZK15WhitePSLog in for availability1 * 1 Un. - 5,94 €