Orders LinkContactUsLinkComponent
Sample bottles, glass
Sample bottles, glass
1000 ml, Sample bottles, GL 45
Catalog # BURK5314-0100
Supplier:   BURKLE
Sample bottles, glass
Catalog # BURK5314-0100
Supplier:  BURKLE
Sample bottles, glass
Sample bottles, glass
1000 ml, Sample bottles, GL 45
Price
26,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Specifications
  • Thread:
    GL 45
  • Capacity:
    1000 ml
  • Description:
    Sample bottles
  • Seq:
    03