Round canisters
2 L, Natural, B53, Standard, 119×119×260 mm
SCTV107711
: S.C.A.T.
SCAT round canisters are ideal for collecting and disposing of smaller quantities of solvents due to their compact dimensions. Several SCAT round canisters can be easily stationed at the HPLC system and equipped with SCAT SafeyWasteCaps.
- Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in the laboratory
- Available in the common threads S65, S70/71 and B83
- Filling capacities range from very compact 2 to 4 L, 10, 25, 50 and 60 L for the collection of voluminous solvents
- Many individual features such as carrying handles, handles or measuring scales, which ensure safe handling of the hazardous substances
SCAT round canisters are a perfect alternative or supplement to conventional SCAT canisters to provide physical separation of solvents and prevent mixing of hazardous liquids.
- Thread:B53
- Type:Standard
- Capacity:2 L
- Colour:Natural
- Seq:01
- W×D×H:119×119×260 mm



