Order Entry
Belgium
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Sep-Pak® Plus

Solid phase extraction cartridges, Sep-Pak® Plus

Sep-Pak® Plus Short, C18, Cartridge, 125 Å, 55 - 105 µm, 0,7 ml, 360 mg
Catalog # WATEWAT020515
Marketsource
Supplier:   WATERS
Solid phase extraction cartridges, Sep-Pak® Plus
Catalog # WATEWAT020515
Supplier:  WATERS
Marketsource
Solid phase extraction cartridges, Sep-Pak® Plus

Sep-Pak® Plus Short, C18, Cartridge, 125 Å, 55 - 105 µm, 0,7 ml, 360 mg

Price
285,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

These cartridges can used with liquid and gas samples and have the unique capability of reverse flow direction, a useful feature for trace enrichment application.

  • Precision design suits to robotic application also more convenient in manual use
  • Sorbents are in short and long body configurations

Can be connected to different devices like filter units, positive flow pumps and syringes, vacuum manifolds, automated sample processor, and in-line reaction systems (PET applications).

Specifications
  • Column Format:
    Cartridge
  • Particle Size:
    55 - 105 µm
  • Pore size:
    125 Å
  • Capacity:
    0,7 ml
  • Description:
    Sep-Pak® Plus Short, C18
  • Seq:
    01
  • Bed weight:
    360 mg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.