TRUCAL ASO 5 LEVEL 5X1ML 1 * 5 Vial
Catalog # DIAS170109910059
Supplier: DIAGNOSTIC SYSTEMS DIASYS
Frequently Bought Together
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Qualitative test paper phosphatesmo MI for phosphatase in milk
Supplier: MACHEREY-NAGEL
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Test kits, photometric, Nitrite, LCK cuvette tests
Supplier: Hach
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test strips, sulphite, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
Supplier: CUSTOM MADE CHEMICALS
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach