Plaster dispenser, QuickFix Elastic
Plaster dispenser with 90 elastic plasters (72×25 mm), 232×33×133 mm
Catalog # 118-0226
Supplier: Plum Safety
Jump to:
Product Details & Documents
Dispenser with elastic finger plasters made from breathable material. The dispenser contains 90 plasters (2 refills) and can be wall-mounted.
- Elastic textile fabric: breathable and skin friendly
- Adapts to movements of the skin and is very comfortable to wear
- Optimum hold even when used in demanding applications
- Dispenser can be operated with just one hand
- Easy to refill
- Very hygienic since each plaster is individually wrapped
Plaster size: 72×25 mm
: Dispenser includes 2×45 plaster strips.
Specifications
- Description:Plaster dispenser with 90 elastic plasters (72×25 mm)
- W×D×H:232×33×133 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



