ContactUsLinkComponent
LIBRA S6 Spectrophotometer supplied with mains lead, starter pack of cuvettes and instruction manual Wavelength Range : 330 800nm Bandwidth : 7nm Accuracy : 2.0nm Photometric Accuracy : 0.01A@1A Resolution : 0.01A Absorbance Range: 0.3 1 * 1 items
Catalog # WAPR80-5000-10
Supplier:  Biochrom
undefined
LIBRA S6 Spectrophotometer supplied with mains lead, starter pack of cuvettes and instruction manual Wavelength Range : 330 800nm Bandwidth : 7nm Accuracy : 2.0nm Photometric Accuracy : 0.01A@1A Resolution : 0.01A Absorbance Range: 0.3 1 * 1 items
Catalog # WAPR80-5000-10
Supplier:  Biochrom
Frequently Bought Together