Order Entry
Belgium
Orders LinkContactUsLinkComponent
Quartz

Quartz

20 kg
Catalog # ROTH4651.7
Marketsource
Supplier:   Roth Carl
Quartz
Catalog # ROTH4651.7
Supplier:  Roth Carl
Marketsource
CAS Number:  14808-60-7
Quartz
Quartz

20 kg

CAS Number:  14808-60-7
Price
Inquire For Price
Log in for availability
1 * 20 kg
Product Details & Documents
Synonyms:  Silicium dioxide, Silicium (IV) oxide, Silicon dioxide, Sand, white quartz
  • CAS Number:14808-60-7
  • Formula:SiO₂
  • MDL Number:MFCD00217788
  • Molecular Weight:60.08 g/mol
  • Boiling Pt:2230 °C (1013 hPa)
  • Melting Pt:1427 °C
  • Density:2.622…2.645 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:238-878-4
Specifications

Specifications

  • Pk:
    20 kg

Specifications