Orders LinkContactUsLinkComponent
VWR®, dNTPs
dNTP set, separate vials of dA, dC, dG, dT, each 100 mM, 4×250 μl, 1 SET
  733-1364
VWR®, dNTPs
  733-1364
 :  
VWR®, dNTPs
VWR®, dNTPs
dNTP set, separate vials of dA, dC, dG, dT, each 100 mM, 4×250 μl, 1 SET
 :  
177,00 €
1 * 1 SET
Ready to use molecular biology grade dNTP mixes and dNTP sets. The dNTP mix is designed to save hands-on time for researchers, and reduce the possibility of contamination by reducing pipetting. dNTP solutions are also available in sets of four individual dNTPs, each 100 mm. Both are convenient for use in DNA polymerisation reactions, DNA labelling and sequencing processes.
  • Available as pre-mixed 10 or 25 mM solutions, or as sets of individual 100 mM dNTP solutions
  • Both pre-mixed and sets have been functionally tested in PCR
  • Purity >99% by HPLC
  • Supplied in solution at pH 7,3 to 7,5
  • Pk:
    1 SET
  • Description:
    dNTP set, separate vials of dA, dC, dG, dT, each 100 mM, 4×250 μl